(2-Hydroxy-4,6-dimethylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A703287-1G A703287-250MG A703287-100MG
77145-802EA 161.26 USD
77145-802 77145-804 77747-407
(2-Hydroxy-4,6-dimethylphenyl)boronic acid 97%
(2-Hydroxy-4,6-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD22414775
CAS Number: 1421935-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR