3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-formylphenylboronic acid pinacol ester

A701843-1G A701843-250MG A701843-5G
77145-786EA 114.58 USD
77145-786 77145-788 77145-790
3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 96%
3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
MDL Number: MFCD18732921
CAS Number: 1352657-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR