1-(3,5-Dichlorophenyl)-2-thiourea 98+%

Supplier: Ambeed
Danger

Synonyms: N-(3,5-Dichlorophenyl)thiourea, 3,5-Dichlorophenylthiourea, 1-(3,5-Dichlorophenyl)thiourea

A699937-1G A699937-25G A699937-5G
77145-766EA 19.1 USD
77145-766 77145-768 77145-770
1-(3,5-Dichlorophenyl)-2-thiourea 98+%
1-(3,5-Dichlorophenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
MDL Number: MFCD00041168
CAS Number: 107707-33-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR