2-Bromo-3,5-difluorotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1,5-difluoro-3-methylbenzene

A696864-10G A696864-1G A696864-250MG A696864-5G
77145-738EA 538.94 USD
77145-738 77145-740 77145-742 77145-744
2-Bromo-3,5-difluorotoluene 95%
2-Bromo-3,5-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.015 g/mol
MDL Number: MFCD28738349
CAS Number: 1807135-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR