(R)-3-(3,4-Dihydroxyphenyl)-2-(((E)-3-(2-((E)-3,4-dihydroxystyryl)-3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid 98%
Supplier: Ambeed
|
Warning
|
A695449-10MG
A695449-1MG
A695449-25MG
A695449-50MG
A695449-5MG
A695449-100MG
A695449-1G
77145-720EA
40.18
USD
77145-720
77145-722
77145-724
77145-726
77145-728
77686-679
77686-680
(R)-3-(3,4-Dihydroxyphenyl)-2-(((E)-3-(2-((E)-3,4-dihydroxystyryl)-3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid 98%
(R)-3-(3,4-Dihydroxyphenyl)-2-(((E)-3-(2-((E)-3,4-dihydroxystyryl)-3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid
Precaución: for Research Use Only
Formula:
C₂₆H₂₂O₁₀ |
MDL Number:
MFCD08458447 CAS Number: 96574-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...