Methyl 5-bromo-3-fluoro-2-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
A694228-100MG
A694228-250MG
77145-714EA
40.32
USD
77145-714
77145-716
Methyl 5-bromo-3-fluoro-2-methylbenzoate 98%
Methyl 5-bromo-3-fluoro-2-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrFO₂ MW: 247.06 g/mol |
MDL Number:
MFCD28739729 CAS Number: 1805501-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...