(1-(tert-Butoxycarbonyl)-1H-pyrrolo[3,2-c]pyridin-2-yl)boronic acid 95%

Supplier: Ambeed
Warning

A693337-250MG A693337-1G
77145-708EA 173.04 USD
77145-708 77747-396
(1-(tert-Butoxycarbonyl)-1H-pyrrolo[3,2-c]pyridin-2-yl)boronic acid 95%
(1-(tert-Butoxycarbonyl)-1H-pyrrolo[3,2-c]pyridin-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₄
MDL Number: MFCD29059780
CAS Number: 1373273-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR