Potassium trifluoro(1-methyl-1H-pyrazol-4-yl)borate 98%
Supplier: Ambeed
|
Warning
|
A693269-1G
A693269-250MG
77145-704EA
182.32
USD
77145-704
77145-706
Potassium trifluoro(1-methyl-1H-pyrazol-4-yl)borate 98%
Potassium trifluoro(1-methyl-1H-pyrazol-4-yl)borate
Precaución: for Research Use Only
Formula:
C₄H₅BF₃KN₂ |
MDL Number:
MFCD09992927 CAS Number: 1365970-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...