tert-Butyl ((2S,3R)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A691852-1G A691852-250MG A691852-5G
77145-692EA 10.31 USD
77145-692 77145-694 77145-696
tert-Butyl ((2S,3R)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate 97%
tert-Butyl ((2S,3R)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD01317802
CAS Number: 162536-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR