3-Bromo-7-chloro-1H-pyrazolo[4,3-c]pyridine 97%
Supplier: Ambeed
|
Danger
|
A691529-1G
A691529-250MG
77145-688EA
190.97
USD
77145-688
77145-690
3-Bromo-7-chloro-1H-pyrazolo[4,3-c]pyridine 97%
3-Bromo-7-chloro-1H-pyrazolo[4,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₆H₃BrClN₃ MW: 232.46 g/mol |
MDL Number:
MFCD22380446 CAS Number: 1357945-65-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...