1-Hydroxy-3,3-dimethylbutan-2-one 97%

Supplier: Ambeed
Danger

A691372-1G A691372-250MG
77145-684EA 345.86 USD
77145-684 77145-686
1-Hydroxy-3,3-dimethylbutan-2-one 97%
1-Hydroxy-3,3-dimethylbutan-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.15 g/mol
MDL Number: MFCD08067721
CAS Number: 38895-88-4
UN: 1224
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR