3-Fluorosalicylaldehyde 97%

Supplier: Ambeed
Warning

A689864-1G A689864-25G A689864-5G
77145-674EA 7.2 USD
77145-674 77145-676 77145-678
3-Fluorosalicylaldehyde 97%
3-Fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO₂
MDL Number: MFCD00003319
CAS Number: 394-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR