1-(4-Methoxybenzyl)-1H-pyrazolo[3,4-b]pyridin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A689556-1G
A689556-250MG
A689556-5G
77145-668EA
67.9
USD
77145-668
77145-670
77145-672
1-(4-Methoxybenzyl)-1H-pyrazolo[3,4-b]pyridin-4-ol 95%
1-(4-Methoxybenzyl)-1H-pyrazolo[3,4-b]pyridin-4-ol
Precaución: for Research Use Only
Formula:
C₁₄H₁₃N₃O₂ MW: 255.27 g/mol |
MDL Number:
MFCD20485436 CAS Number: 924909-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...