Methyl 2-amino-5-chloro-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

A688549-100MG A688549-1G A688549-250MG A688549-5G
77145-646EA 120.95 USD
77145-646 77145-648 77145-650 77145-652
Methyl 2-amino-5-chloro-4-methoxybenzoate 95%
Methyl 2-amino-5-chloro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.63 g/mol
MDL Number: MFCD24626483
CAS Number: 79025-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR