2-Bromo-5-fluoro-4-methylpyrimidine 97%

Supplier: Ambeed
Danger

A686571-100MG A686571-1G A686571-250MG
77145-618EA 36.05 USD
77145-618 77145-620 77145-622
2-Bromo-5-fluoro-4-methylpyrimidine 97%
2-Bromo-5-fluoro-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD28739781
CAS Number: 1805192-57-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR