Benzyl-4-fluorophenyl ketone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Fluorophenyl)-2-phenylethanone

A685647-100G A685647-1G A685647-5G
77145-604EA 315.09 USD
77145-604 77145-606 77145-608
Benzyl-4-fluorophenyl ketone 98%
Benzyl 4-fluorophenyl ketone

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO
MW: 214.2350 g/mol
MDL Number: MFCD00017960
CAS Number: 347-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR