3-Methoxy-1-propanol 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Trimethylene glycol monomethyl ether
A685554-10G
A685554-25G
77145-598EA
11.33
USD
77145-598
77145-600
77145-602
3-Methoxy-1-propanol 97%
3-Methoxy-1-propanol
Precaución: for Research Use Only
Formula:
C₄H₁₀O₂ |
MDL Number:
MFCD00039550 CAS Number: 1589-49-7 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...