5-(Benzyloxy)-7-bromobenzo[d]thiazole 97%
Supplier: Ambeed
|
Warning
|
A685176-1G
A685176-250MG
77145-592EA
335.25
USD
77145-592
77145-594
5-(Benzyloxy)-7-bromobenzo[d]thiazole 97%
5-(Benzyloxy)-7-bromobenzo[d]thiazole
Precaución: for Research Use Only
Formula:
C₁₄H₁₀BrNOS MW: 320.20 g/mol |
MDL Number:
MFCD29922322 CAS Number: 1650547-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...