Cyclohexanemethylamine 98%

Supplier: Ambeed
Danger

Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

A684885-25G A684885-5G
77145-588EA 44.29 USD
77145-588 77145-590
Cyclohexanemethylamine 98%
Cyclohexanemethylamine

Precaución: for Research Use Only

Formula: C₇H₁₅N
MDL Number: MFCD00001520
CAS Number: 3218-02-8
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR