4,5-Diiodo-2-methylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Diiodo-2-methyl-1H-imidazole

A683780-1G A683780-250MG A683780-25G A683780-5G
77145-580EA 19.57 USD
77145-580 77145-582 77145-584 77145-586
4,5-Diiodo-2-methylimidazole 98%
4,5-Diiodo-2-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₄I₂N₂
MW: 333.9 g/mol
MDL Number: MFCD00464337
CAS Number: 73746-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR