Silver(I) acetylacetonate 98%
Supplier: Ambeed
|
Warning
|
A683739-250MG
A683739-5G
A683739-1G
77145-576EA
18.55
USD
77145-576
77145-578
77686-659
Silver(I) acetylacetonate 98%
Silver(I) acetylacetonate
Precaución: for Research Use Only
Formula:
C₅H₈AgO₂ |
MDL Number:
MFCD00000012 CAS Number: 15525-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...