Methyl 6-hydroxy-5-iodonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-hydroxy-5-iodopyridine-3-carboxylate, 2-Hydroxy-3-iodo-5-(methoxycarbonyl)pyridine

A681808-100MG A681808-1G A681808-250MG A681808-5G
77145-542EA 35.01 USD
77145-542 77145-544 77145-546 77145-548
Methyl 6-hydroxy-5-iodonicotinate 98%
Methyl 6-hydroxy-5-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₆INO₃
MW: 279.04 g/mol
MDL Number: MFCD17168292
CAS Number: 885950-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR