Sulforhodamine 101 97%

Supplier: Ambeed
Warning

Synonyms: Sulforhodamine 101, Texas Red, SR 101, Sulforhodamine 640, 9-(2,4-Disulphophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium inner salt

A678873-100MG A678873-50MG
77145-536EA 117.76 USD
77145-536 77617-824
Sulforhodamine 101 97%
Sulforhodamine 101

Precaución: for Research Use Only

Formula: C₃₁H₃₀N₂O₇S₂
MW: 606.708 g/mol
MDL Number: MFCD00012407
CAS Number: 60311-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR