2,4-Dibromo-6-fluoropyridine 97%

Supplier: Ambeed
Warning

A676559-100MG A676559-250MG
77145-522EA 114.58 USD
77145-522 77145-524
2,4-Dibromo-6-fluoropyridine 97%
2,4-Dibromo-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂FN
MW: 254.88 g/mol
MDL Number: MFCD28055251
CAS Number: 1622844-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR