(4-Chlorophenylthio)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A676035-1G
A676035-250MG
A676035-5G
77145-516EA
18.54
USD
77145-516
77145-518
77145-520
(4-Chlorophenylthio)acetic acid 98%
(4-Chlorophenylthio)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂S |
MDL Number:
MFCD00021762 CAS Number: 3405-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...