4-Bromo-2-formylbenzoic acid 98%

Supplier: Ambeed
Warning

A673972-100MG A673972-1G A673972-250MG
77145-510EA 59.42 USD
77145-510 77145-512 77145-514
4-Bromo-2-formylbenzoic acid 98%
4-Bromo-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.027 g/mol
MDL Number: MFCD11036374
CAS Number: 871502-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR