Methyl-5-(chloromethyl)-2-furoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 5-(chloromethyl)-2-furancarboxylate, Methyl 5-(chloromethyl)furan-2-carboxylate

A673943-10G A673943-1G A673943-5G
77145-504EA 63.66 USD
77145-504 77145-506 77145-508
Methyl-5-(chloromethyl)-2-furoate 98%
Methyl-5-(chloromethyl)-2-furoate

Precaución: for Research Use Only

Formula: C₇H₇ClO₃
MW: 174.58 g/mol
MDL Number: MFCD00020922
CAS Number: 2144-37-8
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR