4-Cyano-3-ethoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A673507-1G A673507-250MG A673507-5G
77145-498EA 194.66 USD
77145-498 77145-500 77145-502
4-Cyano-3-ethoxyphenylboronic acid 97%
4-Cyano-3-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₃
MDL Number: MFCD19684099
CAS Number: 2096338-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR