Pentafluoropyridine 98%

Supplier: Ambeed
Danger

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

A672533-25G A672533-5G A672533-1G
77145-492EA 42.44 USD
77145-492 77145-494 77617-810
Pentafluoropyridine 98%
Pentafluoropyridine

Precaución: for Research Use Only

Formula: C₅F₅N
MW: 169.05 g/mol
MDL Number: MFCD00006225
CAS Number: 700-16-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR