2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98%

Supplier: Ambeed
Danger

A670624-1G A670624-250MG
77145-488EA 39.26 USD
77145-488 77145-490
2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98%
2-(Bromomethyl)-1-fluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD00042918
CAS Number: 1958-93-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR