3-(Bromomethyl)-2-methyl-1,1'-biphenyl 97%

Supplier: Ambeed
Danger

A669446-250MG A669446-1G
77145-474EA 142.17 USD
77145-474 77552-036
3-(Bromomethyl)-2-methyl-1,1'-biphenyl 97%
3-(Bromomethyl)-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.15 g/mol
MDL Number: MFCD29923197
CAS Number: 116175-22-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR