3-Borono-4,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A668921-1G A668921-250MG
77145-460EA 162.32 USD
77145-460 77145-462
3-Borono-4,5-difluorobenzoic acid 97%
3-Borono-4,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
MDL Number: MFCD28127178
CAS Number: 2377605-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR