2,6-Dichloro-4-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A667876-1G A667876-5G
77145-454EA 172.93 USD
77145-454 77573-152
2,6-Dichloro-4-nitrobenzoic acid 97%
2,6-Dichloro-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₄
MW: 236.00 g/mol
MDL Number: MFCD09999836
CAS Number: 22509-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR