(R)-3-Hydroxy-2-(methylamino)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-D-Me-Ser-OH

A667309-100MG A667309-250MG
77145-444EA 59.74 USD
77145-444 77145-446
(R)-3-Hydroxy-2-(methylamino)propanoic acid 97%
(R)-3-Hydroxy-2-(methylamino)propanoic acid

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD19220663
CAS Number: 915405-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR