(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-(-)-Bis(diphenylphosphino)butane, (2S,3S)-Butane-2,3-diylbis(diphenylphosphine), (S,S)-CHIRAPHOS

A666060-100MG A666060-1G A666060-250MG A666060-5G
77145-436EA 38.2 USD
77145-436 77145-438 77145-440 77145-442
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥98%, ee 99%
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane

Precaución: for Research Use Only

Formula: C₂₈H₂₈P₂
MW: 426.47 g/mol
MDL Number: MFCD00009836
CAS Number: 64896-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR