(R)-(-)-1-Amino-1-phenyl-2-methoxyethane 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Methoxy-1-phenylethanamine

A665312-100MG A665312-1G A665312-250MG
77145-430EA 64.72 USD
77145-430 77145-432 77145-434
(R)-(-)-1-Amino-1-phenyl-2-methoxyethane 98%
(R)-(-)-1-Amino-1-phenyl-2-methoxyethane

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.206 g/mol
MDL Number: MFCD01862274
CAS Number: 64715-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR