4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%

Supplier: Ambeed
Warning

Synonyms: [2-(Morpholin-4-yl)pyridin-4-yl]methanol, (2-Morpholinopyridin-4-yl)methanol, 4-(Hydroxymethyl)-2-(morpholin-4-yl)pyridine

A664656-100MG A664656-1G A664656-250MG
77145-418EA 33.95 USD
77145-418 77145-420 77145-422
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
MDL Number: MFCD08271915
CAS Number: 556109-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR