Ferrozine 98% (contains < 2% H₂O)

Supplier: Ambeed
Warning

Synonyms: 4,4'-(3-(Pyridin-2-yl)-1,2,4-triazine-5,6-diyl)dibenzenesulphonic acid monosodium salt, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulphonic acid monosodium salt

A664488-1G A664488-250MG
77145-410EA 30.91 USD
77145-410 77686-632
Ferrozine 98% (contains < 2% H₂O)
Ferrozine

Precaución: for Research Use Only

Formula: C₂₀H₁₃N₄NaO₆S₂
MW: 492.46 g/mol
MDL Number: MFCD00078417
CAS Number: 69898-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR