4-Methoxy-4'-methyldiphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-N-(p-tolyl)aniline, N-(4-Methoxyphenyl)-p-tolylamine

A664127-1G A664127-250MG A664127-25G A664127-5G
77145-402EA 22.66 USD
77145-402 77145-404 77145-406 77145-408
4-Methoxy-4'-methyldiphenylamine 98%
4-Methoxy-4'-methyldiphenylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
MDL Number: MFCD00092922
CAS Number: 39253-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR