tert-Butyl (4-(piperidin-4-yloxy)phenyl)carbamate 95%

Supplier: Ambeed
Warning

A663324-1G A663324-250MG
77145-392EA 264.17 USD
77145-392 77145-394
tert-Butyl (4-(piperidin-4-yloxy)phenyl)carbamate 95%
tert-Butyl (4-(piperidin-4-yloxy)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₃
MW: 292.37 g/mol
MDL Number: MFCD08460606
CAS Number: 1820604-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR