1-Butyl-3-methylimidazolium hydrogen sulphate 97%

Supplier: Ambeed
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium hydrogensulphate

A662199-5G A662199-25G
77145-384EA 18.54 USD
77145-384 77595-008
1-Butyl-3-methylimidazolium hydrogen sulphate 97%
1-Butyl-3-methylimidazolium hydrogen sulphate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄S
MW: 236.29
MDL Number: MFCD06798197
CAS Number: 262297-13-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR