4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)phthalonitrile 99%

Supplier: Ambeed
Danger

A662061-100MG A662061-1G A662061-250MG
77145-378EA 45.62 USD
77145-378 77145-380 77145-382
4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)phthalonitrile 99%
4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)phthalonitrile

Precaución: for Research Use Only

Formula: C₁₅H₉BN₂O₃
MW: 276.05 g/mol
MDL Number: MFCD18633196
CAS Number: 906673-33-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR