1-(2,5-Dimethoxyphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 97%

Supplier: Ambeed
Warning

A661346-1G A661346-5G
77145-374EA 201.58 USD
77145-374 77145-376
1-(2,5-Dimethoxyphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 97%
1-(2,5-Dimethoxyphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea

Precaución: for Research Use Only

Formula: C₂₁H₂₇BN₂O₅
MW: 398.26 g/mol
MDL Number: MFCD20231485
CAS Number: 2096997-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR