(R)-4-(tert-Butoxy)-2-ethyl-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A659645-250MG A659645-1G
77145-364EA 123.07 USD
77145-364 77308-076
(R)-4-(tert-Butoxy)-2-ethyl-4-oxobutanoic acid 97%
(R)-4-(tert-Butoxy)-2-ethyl-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
MDL Number: MFCD15144617
CAS Number: 210048-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR