(R)-tert-Butyl-2-benzylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: (R)-tert-Butyl 2-benzylpiperazine-1-carboxylate

A658908-100MG A658908-250MG A658908-5G A658908-1G
77145-356EA 16.98 USD
77145-356 77145-358 77145-360 77573-126
(R)-tert-Butyl-2-benzylpiperazine-1-carboxylate 97%
(R)-tert-Butyl-2-benzylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
MDL Number: MFCD07772094
CAS Number: 947684-78-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR