4-(Bromomethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)benzene-1-sulphonyl chloride

A657725-1G A657725-5G A657725-25G
77145-350EA 7.43 USD
77145-350 77145-352 77552-006
4-(Bromomethyl)benzenesulfonyl chloride 98%
4-(Bromomethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
MDL Number: MFCD00156129
CAS Number: 66176-39-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR