4-(Trifluoromethyl)thiobenzamide 98%

Supplier: Ambeed
Danger

A655737-100G A655737-10G A655737-25G A655737-5G
77145-336EA 102.91 USD
77145-336 77145-338 77145-340 77145-342
4-(Trifluoromethyl)thiobenzamide 98%
4-(Trifluoromethyl)thiobenzamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NS
MW: 205.2 g/mol
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR