1,2,4-Triethoxybenzene 97%

Supplier: Ambeed
Warning

A654746-100G A654746-25G A654746-5G
77145-328EA 133.91 USD
77145-328 77145-330 77145-332
1,2,4-Triethoxybenzene 97%
1,2,4-Triethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₃
MDL Number: MFCD00026769
CAS Number: 41827-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR