2-Bromo-1-(2-fluoropyridin-4-yl)ethanone 97%

Supplier: Ambeed
Danger

A653383-100MG A653383-250MG
77145-316EA 189.52 USD
77145-316 77145-318
2-Bromo-1-(2-fluoropyridin-4-yl)ethanone 97%
2-Bromo-1-(2-fluoropyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MDL Number: MFCD18910113
CAS Number: 1202854-31-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR