Ethyl 7-amino-5-chloro-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A644870-100MG A644870-250MG
77145-286EA 175.05 USD
77145-286 77145-288
Ethyl 7-amino-5-chloro-1H-indole-2-carboxylate 97%
Ethyl 7-amino-5-chloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
MDL Number: MFCD20923225
CAS Number: 59694-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR